Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1640

Sigma-Aldrich

DC_AC50

≥98% (HPLC)

Synonym(s):

3-Amino-N-(2-bromo-4,6-difluorophenyl)-6,7-dihydro-5H-cyclopenta[b]thieno[3,2-e]pyridine-2-carboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$69.00
25 MG
$279.00

$69.00

List Price$138.00Save 50%
Web-Only Promotion

Available to ship onMay 05, 2025Details


Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
5 MG
$69.00
25 MG
$279.00

About This Item

Empirical Formula (Hill Notation):
C17H12BrF2N3OS
CAS Number:
Molecular Weight:
424.26
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$69.00

List Price$138.00Save 50%
Web-Only Promotion

Available to ship onMay 05, 2025Details


Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.

Quality Level

assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

NC1=C(C(NC2=C(Br)C=C(F)C=C2F)=O)SC3=NC4=C(CCC4)C=C31

InChI

1S/C17H12BrF2N3OS/c18-10-5-8(19)6-11(20)14(10)23-16(24)15-13(21)9-4-7-2-1-3-12(7)22-17(9)25-15/h4-6H,1-3,21H2,(H,23,24)

InChI key

DFNOJNBNTVQPCA-UHFFFAOYSA-N

Biochem/physiol Actions

DC_AC50 is an inhibitor of copper trafficking proteins Atox1 and CCS, resulting in a disruption of cellular copper transport. Cancer cells rely more heavily on Atox1 and CCS than normal cells. DC_AC50 blockade of copper trafficking induces cellular oxidative stress, affecting cancer cells more strongly than normal cells, and leading to the inhibition of cancer cell proliferation.
DC_AC50 is an inhibitor of copper trafficking proteins Atox1 and CCS.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service