Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1632

Sigma-Aldrich

Ceftezole Sodium

≥90% (HPLC)

Synonym(s):

(6R,7R)-8-oxo-7-{[2-(tetrazol-1-yl)acetyl]amino}-3-(1,3,4-thiadiazol-2-ylsulfanylmethyl)-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid sodium salt, Alomen, Celoslin, Falomesin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H11N8O4S3 · Na
CAS Number:
Molecular Weight:
462.46
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic

assay

≥90% (HPLC)

form

powder

solubility

H2O: soluble 200 mg/L

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C1[C@@H](NC(CN2N=NN=C2)=O)[C@]3([H])N1C(C([O-])=O)=C(CSC4=NN=CS4)CC3.[Na+]

InChI

1S/C13H12N8O4S3.Na/c22-7(1-20-4-14-18-19-20)16-8-10(23)21-9(12(24)25)6(2-26-11(8)21)3-27-13-17-15-5-28-13;/h4-5,8,11H,1-3H2,(H,16,22)(H,24,25);/q;+1/p-1/t8-,11-;/m1./s1

InChI key

UGUMHWUOXWFPFH-JHQAJZDGSA-M

Biochem/physiol Actions

Ceftezole is a first generation cephalosporin with antibacterial activity.
Ceftezole is a first generation cephalosporin with antibacterial activity. Ceftezole binds to and inactivates penicillin-binding proteins (PBPs) located on the inner membrane of the bacterial cell wall. Inactivation of PBPs interferes with the cross-linkage of peptidoglycan chains necessary for bacterial cell wall strength and rigidity causing bacterial cell lysis.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service