Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1631

Sigma-Aldrich

Cefodizime Sodium

≥96% (HPLC)

Synonym(s):

5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[[(2Z)-(2-amino-4-thiazoly)(methoxyimino)acetyl}-3-[[[5-(carboxymethyl)-4-methyl-2-thiazilyl]-8-oxo-, sodium salt (1:2), (6R,7R)-

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H18N6O7S4 · 2Na
CAS Number:
Molecular Weight:
628.63
UNSPSC Code:
12352200
PubChem Substance ID:

biological source

synthetic

assay

≥96% (HPLC)

form

powder

solubility

H2O: soluble 270 mg/mL

storage temp.

2-8°C

SMILES string

O=C1[C@@H](NC(/C(C2=CSC(N)=N2)=N\OC)=O)[C@]3([H])N1C(C([O-])=O)=C(CSC4=NC(C)=C(CC([O-])=O)S4)CC3

InChI

1S/C20H20N6O7S4.2Na/c1-7-10(3-11(27)28)37-20(22-7)36-5-8-4-34-17-13(16(30)26(17)14(8)18(31)32)24-15(29)12(25-33-2)9-6-35-19(21)23-9;;/h6,13,17H,3-5H2,1-2H3,(H2,21,23)(H,24,29)(H,27,28)(H,31,32);;/q;2*+1/p-2/b25-12-;;/t13-,17-;;/m1../s1

InChI key

WBOBLQIRACJNPA-AEKYOGSZSA-L

Biochem/physiol Actions

Cefodizime is a third generation cephalosporin with a broad spectrum of antibacterial activity.
Cefodizime is a third generation cephalosporin with a broad spectrum of antibacterial activity. Cefodizime was demonstrated to be effective against upper and lower respiratory tract infections, urinary tract infections, and gonorrhea. Cefodizime regulates the cytokines expression of neutrophils through toll-like receptor 4 (TLR-4) stimulated by Klebsiella pneumonie.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service