Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1622

Sigma-Aldrich

iKIX1

≥98% (HPLC)

Synonym(s):

2-(2-Cyanoacetyl)-N-(3,4-dichlorophenyl)hydrazine-1-carbothioamide, 3-[(2-Cyanoacetyl)amino]-1-(3,4-dichlorophenyl)thiourea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8Cl2N4OS
CAS Number:
Molecular Weight:
303.17
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 25 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NNC(NC1=CC(Cl)=C(C=C1)Cl)=S)CC#N

InChI

1S/C10H8Cl2N4OS/c11-7-2-1-6(5-8(7)12)14-10(18)16-15-9(17)3-4-13/h1-2,5H,3H2,(H,15,17)(H2,14,16,18)

InChI key

XKQJVBSDCOCZFV-UHFFFAOYSA-N

Biochem/physiol Actions

iKIX1 is an antifungal agent that can re-sensitize drug-resistant Candida glabrata to azole antifungals. C. glabrata fungal infections that are resistant to azoles are an increasing problem. Drug-resistant C. glabrata most commonly contain point mutations in Pdr1 that render it active. iKIX1 acts by inhibiting the protein–protein interaction between the fungal transcription factor Pdr1 and the KIX domain of the MED15 subunit of the co-activator Mediator, thus inhibiting Pdr1-dependent gene activation of key regulators of the multidrug resistance pathway. iKIX1 reduced efflux pump gene expression and restored azole-sensitivity to PDR1 gain-of-function mutant Candida glabrata strains in plate assays. In mouse assays, combination treatment with iKIX1 and fluconazole showed significant reduction of fungal burden in mice injected with two different azole-resistant C. glabrata strains.
iKIX1 is an antifungal inhibitor of a protein-protein interaction between the fungal transcription factor Pdr1 and the KIX domain of the MED15 subunit of the co-activator Mediator.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service