Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1598

Sigma-Aldrich

Bis(heptyl)-cognitin dihydrochloride

≥98% (HPLC)

Synonym(s):

9-Amino-1,2,3,4-tetrahydroacridine bis 1,7-heptylene dihydrochloride, B7C, Bis(7)-cognitin dihydrochloride, Heptylenebis-tacrine dihydrochloride, N,N′-Bis-(1,2,3,4-tetrahydro-acridin-9-yl)-heptane-1,7-diamine dihydrochloride, heptylene-bis(THA) dihydrochloride, heptylene-bis(tacrine) dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H40N4 · 2HCl
CAS Number:
Molecular Weight:
565.62
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.25

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

Cl.Cl.C(CCCNc1c2CCCCc2nc3ccccc13)CCCNc4c5CCCCc5nc6ccccc46

InChI

1S/C33H40N4.2ClH/c1(2-12-22-34-32-24-14-4-8-18-28(24)36-29-19-9-5-15-25(29)32)3-13-23-35-33-26-16-6-10-20-30(26)37-31-21-11-7-17-27(31)33;;/h4,6,8,10,14,16,18,20H,1-3,5,7,9,11-13,15,17,19,21-23H2,(H,34,36)(H,35,37);2*1H

InChI key

RHKXINFBJWDTSK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Bis(heptyl)-cognitin (B7C) is a potent neuroprotective agent and cognitive enhancer. It is thought to have multiple mechanisms of action including inhibition of AChE, and inihbititon of the amyloid precursor protein/beta-amyloid cascade. It has been shown to inhibit the formation of Aβ1-42 oligomers and reduce the amount of pre-formed Aβ1-42 oligomers, and to induce robust neurite outgrowth in PC12 cells.[1]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lan Chang et al.
Scientific reports, 5, 10256-10256 (2015-07-22)
β-amyloid (Aβ) oligomers have been closely implicated in the pathogenesis of Alzheimer's disease (AD). We found, for the first time, that bis(heptyl)-cognitin, a novel dimeric acetylcholinesterase (AChE) inhibitor derived from tacrine, prevented Aβ oligomers-induced inhibition of long-term potentiation (LTP) at
Ren-wen Han et al.
Brain research, 1470, 59-68 (2012-07-04)
Our previous reports indicated that bis(propyl)-cognitin (B3C) and bis(heptyl)-cognitin (B7C), as novel dimers derived from tacrine, may be potential multifunctional drugs for treating Alzheimer's disease. There is little knowledge on the cognitive function of B3C while B7C appeared to reverse
Sheng-Quan Hu et al.
CNS neuroscience & therapeutics, 21(6), 520-529 (2015-04-29)
Neurodegenerative disorders are caused by progressive neuronal loss in the brain, and hence, compounds that could promote neuritogenesis may have therapeutic values. In this study, the effects of bis(heptyl)-cognitin (B7C), a multifunctional dimer, on neurite outgrowth were investigated in both

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service