Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1597

Sigma-Aldrich

SRI-29574

≥98% (HPLC)

Synonym(s):

N-(2,2-Diphenylethyl)-2-(imidazo[1,2-a]pyridin-6-yl)quinazolin-4-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H23N5
Molecular Weight:
441.53
UNSPSC Code:
12352202
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

Biochem/physiol Actions

DAT is crucial for regulating the levels of DA (dopamine). It is also responsible for the reuptake of 1-methyl-4-phenylpyridinium (MPP+), neurotoxin which mediates symptoms similar to Parkinson′s disease.[1] Changes in DAT activity is associated with attention-deficit/hyperactivity and autism spectrum disorders.[2]
SRI-29574 is a potent DAT (dopamine transporter) allosteric modulator that partially inhibits DAT uptake without affecting binding to DAT.
SRI-29574 is a potent DAT (dopamine transporter) allosteric modulator that partially inhibits DAT uptake without affecting binding to DAT. SRI-29574 is a noncompetitive inhibitor of [3H]DA uptake. Also SRI-29574 partially inhibits SERT and NET uptake.

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sijia Wu et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(50), 15480-15485 (2015-12-02)
The dopamine (DA) transporter (DAT) facilitates high-affinity presynaptic DA reuptake that temporally and spatially constrains DA neurotransmission. Aberrant DAT function is implicated in attention-deficit/hyperactivity disorder and autism spectrum disorder. DAT is a major psychostimulant target, and psychostimulant reward strictly requires
Beryl Luk et al.
PloS one, 10(8), e0136641-e0136641 (2015-08-26)
The regulation of the dopamine transporter (DAT) impacts extracellular dopamine levels after release from dopaminergic neurons. Furthermore, a variety of protein partners have been identified that can interact with and modulate DAT function. In this study we show that DJ-1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service