Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1592

Sigma-Aldrich

Pimprinine

≥98% (HPLC)

Synonym(s):

3-(2-Methyl-5-oxazolyl)-1H-indole, APHE 3, NSC 80793, WS 30581C

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$428.00

$428.00


Available to ship onMay 02, 2025Details


Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
25 MG
$428.00

About This Item

Empirical Formula (Hill Notation):
C12H10N2O
CAS Number:
Molecular Weight:
198.22
UNSPSC Code:
12352200
NACRES:
NA.77

$428.00


Available to ship onMay 02, 2025Details


Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.

Quality Level

assay

≥98% (HPLC)

form

powder

color

white to brown

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

[nH]1c2c(c(c1)c3[o]c(nc3)C)cccc2

InChI

1S/C12H10N2O/c1-8-13-7-12(15-8)10-6-14-11-5-3-2-4-9(10)11/h2-7,14H,1H3

InChI key

WZJPGCHCOHYLMB-UHFFFAOYSA-N

Biochem/physiol Actions

Pimprinine is an indole alkaloid extracted from Streptomyces species that exhibits anticonvulsant activity. Pimprinine is a potent MAO inhibitor. Also, Pimprinine effectively inhibits replication of enterovirus 71 (EV71) and adenovirus 7 (ADV-7).
Pimprinine is an indole alkaloid that exhibits anticonvulsant activity.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service