Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1587

Sigma-Aldrich

Perlapine

≥98% (HPLC)

Synonym(s):

6-(4-Methyl-1-piperazinyl)-11H-dibenz[b,e]azepine, 6-(4-Methyl-1-piperazinyl)morphanthridine, CID 16106

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21N3
CAS Number:
Molecular Weight:
291.39
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

room temp

SMILES string

CC(C)(C1=CC=C(CN2CCN(C(C3=CC=C(Cl)C=C3)C4=CC=CC=C4)CC2)C=C1)C.[H]Cl.[H]Cl

InChI

1S/C28H33ClN2.2ClH/c1-28(2,3)25-13-9-22(10-14-25)21-30-17-19-31(20-18-30)27(23-7-5-4-6-8-23)24-11-15-26(29)16-12-24;;/h4-16,27H,17-21H2,1-3H3;2*1H

InChI key

SDBHDSZKNVDKNU-UHFFFAOYSA-N

Gene Information

human ... HRH1(3269)

Biochem/physiol Actions

Perlapine is a potent and selective agonist of hM3Dq DREADD (Designer Receptors Exclusively Activated by Designer Drugs), which does not activate the native human M3 receptor. Perlapine is a neuroleptic agent that exhibits sedative and sleep-promoting properties.
Potent and selective agonist of hM3Dq DREADD

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service