SML1585
ML116
≥95% (HPLC)
Synonym(s):
4-Benzyl-1-{thieno[2,3-d]pyrimidin-4-yl}piperidine, 4-[4-(Phenylmethyl)-1-piperidinyl]-thieno[2,3-d]pyrimidine, CID 2100018
About This Item
Recommended Products
assay
≥95% (HPLC)
form
powder
color
white to brown
solubility
DMSO: 5 mg/mL, clear (warmed)
storage temp.
room temp
InChI
1S/C18H19N3S/c1-2-4-14(5-3-1)12-15-6-9-21(10-7-15)17-16-8-11-22-18(16)20-13-19-17/h1-5,8,11,13,15H,6-7,9-10,12H2
InChI key
AJQCKNBBHOVJGO-UHFFFAOYSA-N
General description
Biochem/physiol Actions
signalword
Warning
hcodes
pcodes
Hazard Classifications
Acute Tox. 4 Oral
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service