Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1580

Sigma-Aldrich

NSC156529

≥98% (HPLC)

Synonym(s):

6-Methyl-2-phenyl-10-(phenylthio)-1,2-dihydro-24-anthra[1,9-bc]thiophene chloride, 6-Methyl-2-phenyl-10-(phenylthio)-1H-anthra[1,9-bc]thiophenium chloride, CID 3246498, NSC 156529, NSC-156529

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H21ClS2
CAS Number:
Molecular Weight:
457.05
UNSPSC Code:
12352202
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

InChI

1S/C28H21S2.ClH/c1-19-22-14-8-16-25(29-20-10-4-2-5-11-20)27(22)24-18-30(21-12-6-3-7-13-21)26-17-9-15-23(19)28(24)26;/h2-17H,18H2,1H3;1H/q+1;/p-1

InChI key

KLQVNXJBZZWIIV-UHFFFAOYSA-M

Biochem/physiol Actions

NSC156529 is a potent inhibitor of Akt/PKB pathway that decreases the proliferation of human cancer cells in vitro, and inhibits tumor growth in a prostate tumor xenograft model. NSC156529 inhibits interaction of PDPK1 and AKT1 by direct binding to PDPK1. NSC156529 increases the differentiation status of cancer cells.
Potent inhibitor of Akt/PKB pathway

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service