SML1574
Sarcophine
≥97% (HPLC)
Synonym(s):
(+)-Sarcophine, (2S,7S,8S)-Sarcophine, 5,8,12-Tetramethyl-oxireno[9,10]cyclotetradeca[1,2-b]furan-9(1aH)-one, NSC 250434, Sarcophin, [1aS-(1aR*,4E,10aR*,11E,14aR*)]-2,3,6,7,10a,13,14,14a-Octahydro-1a,5,8,12-tetramethyl-oxireno[9,10]cyclotetradeca[1,2-b]furan-9(1aH)-one
About This Item
Recommended Products
biological source
(isolated from soft coral Sarcophyton glaucum)
assay
≥97% (HPLC)
form
powder
color
white to beige
storage temp.
2-8°C
InChI
1S/C20H28O3/c1-13-6-5-11-20(4)18(23-20)10-8-14(2)12-17-16(9-7-13)15(3)19(21)22-17/h6,12,17-18H,5,7-11H2,1-4H3/b13-6+,14-12+
InChI key
CGAKBBMRMLAYMY-VRDMJVMJSA-N
Biochem/physiol Actions
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service