Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1568

Sigma-Aldrich

ZINC05626394

≥98% (HPLC)

Synonym(s):

2,3-Dihydro-6-[(phenylthio)methyl]-2-thioxo-4(1H)-pyrimidinone

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$94.40

$94.40


Available to ship onApril 29, 2025Details


Request a Bulk Order

Select a Size

Change View
5 MG
$94.40

About This Item

Empirical Formula (Hill Notation):
C11H10N2OS2
CAS Number:
Molecular Weight:
250.34
UNSPSC Code:
12352200
NACRES:
NA.77

$94.40


Available to ship onApril 29, 2025Details


Request a Bulk Order

Quality Level

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 25 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

ZINC05626394 is a derivative of propylthiouracil. It is a cell penetrant and a potent inhibitor of CYB5R3 (NADH cytochrome b5 reductase 3).

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service