Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1536

Sigma-Aldrich

(-)-p-Bromolevamisole oxalate Ready Made Solution

0.8 mg/mL in DMSO

Synonym(s):

(-)-4-Bromotetramisole oxalate, 6-Bromolevamisole oxalate, S(-)-6-(4-Bromophenyl)-2,3,5,6-tetrahydroimidazo[2,1-b]thiazole oxalate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11BrN2S · C2H2O4
CAS Number:
Molecular Weight:
373.22
EC Number:
UNSPSC Code:
12352000

form

liquid

Quality Level

concentration

0.8 mg/mL in DMSO

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C11H11BrN2S.C2H2O4/c12-9-3-1-8(2-4-9)10-7-14-5-6-15-11(14)13-10;3-1(4)2(5)6/h1-4,10H,5-7H2;(H,3,4)(H,5,6)

InChI key

ZULBIBHDIQCNIS-UHFFFAOYSA-N

Biochem/physiol Actions

(-)-p-Bromolevamisole oxalate inhibits L-isoforms of alkaline phosphatases.

Other Notes

Stable for at least two years at 2-8° C.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

188.6 °F

flash_point_c

87 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service