Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1494

Sigma-Aldrich

BMS-599626

≥98% (HPLC)

Synonym(s):

(3S)-3-Morpholinylmethyl-[4-[[1-[(3-fluorophenyl)methyl]-1H-indazol-5-yl]amino]-5-methylpyrrolo[2,1-f][1,2,4]triazin-6-yl]-Carbamic acid, AC480

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H27FN8O3
CAS Number:
Molecular Weight:
530.55
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

CC(C(NC(OC[C@@H]1COCCN1)=O)=C2)=C3N2N=CN=C3NC4=CC(C=NN5CC6=CC(F)=CC=C6)=C5C=C4

InChI

1S/C27H27FN8O3/c1-17-23(34-27(37)39-15-22-14-38-8-7-29-22)13-36-25(17)26(30-16-32-36)33-21-5-6-24-19(10-21)11-31-35(24)12-18-3-2-4-20(28)9-18/h2-6,9-11,13,16,22,29H,7-8,12,14-15H2,1H3,(H,34,37)(H,30,32,33)/t22-/m0/s1

InChI key

LUJZZYWHBDHDQX-QFIPXVFZSA-N

Gene Information

Biochem/physiol Actions

BMS-599626 is a potent inhbitor of Pan-HER kinase.
BMS-599626 is a potent inhbitor of human epidermal growth factor receptors 1 and 2 (HER1 and HER2, IC50 = 20 and 30 nM, respectively). BMS599626 inhibits proliferation of several HER-expressing cancer cell lines, including HER2-overexpressing breast cancer lines, but does not affect the proliferation rate of HER-negative cancer cell lines.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service