Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1472

Sigma-Aldrich

TNIIIA2 trifluoroacetate salt

≥95% (HPLC)

Synonym(s):

L-Arginyl-L-seryl-L-threonyl-L-α-aspartyl-L-leucyl-L-prolylglycyl-L-leucyl-L-lysyl-L-alanyl-L-alanyl-L-threonyl-L-histidyl-L-tyrosyl-L-threonyl-L-isoleucyl-L-threonyl-L-isoleucyl-L-arginylglycyl-L-valyl-L-cysteine trifluoroacetate salt, RSTDLPGLKAATHYTITIRGVC trifluoroacetate salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$274.00

$274.00


Check Cart for Availability

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
1 MG
$274.00

About This Item

Empirical Formula (Hill Notation):
C103H173N31O31S · xC2HF3O2
CAS Number:
Molecular Weight:
2373.73 (free base basis)
UNSPSC Code:
41106514
NACRES:
NA.77

$274.00


Check Cart for Availability

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.

Quality Level

assay

≥95% (HPLC)

form

film

storage condition

desiccated

color

colorless

shipped in

wet ice

storage temp.

−20°C

Biochem/physiol Actions

TNIIIA2 is a Tenascin-C driven proadhesive peptide that potently activates β1-integrins including α5β1. TNIIIA2 forces hematopoietic tumor cell lines to adhere via VLA-4 to fibronectin, which induces apoptosis. It appears that TNIIIA2 uses Syndecan-4 as a receptor.
TNIIIA2 is a potent activator of β1-integrins including α5β1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service