Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1436

Sigma-Aldrich

KS 370G

≥98% (HPLC)

Synonym(s):

3-(3,4-Dihydroxyphenyl)-N-(2-phenylethyl)-2-propenamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H17NO3
CAS Number:
Molecular Weight:
283.32
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(/C=C/C1=CC=C(O)C(O)=C1)NCCC2=CC=CC=C2

InChI

1S/C17H17NO3/c19-15-8-6-14(12-16(15)20)7-9-17(21)18-11-10-13-4-2-1-3-5-13/h1-9,12,19-20H,10-11H2,(H,18,21)/b9-7+

InChI key

QOWABIXYAFJMQE-VQHVLOKHSA-N

Biochem/physiol Actions

KS370G is a synthetic caffeamide derivative that exhibits hypoglycemic effects in mice model of diabetes.
KS370G is a synthetic caffeamide derivative that exhibits hypoglycemic effects in mice model of diabetes. Recent study indicates that KS370G exhibits antifibrotic properties in the murine unilateral ureteral obstruction (UUO) model by reduction of inflammation and oxidative stress.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service