SML1425
45-54W trifluoroacetate salt
≥95% (HPLC)
Synonym(s):
Ac-KDGIVNGVKA-NH2 trifluoroacetate salt, Ac-Lys-Asp-Gly-Ile-Val-Asn-Gly-Val-Lys-Ala-NH2 trifluoroacetate salt
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C45H80N14O14 · xC2HF3O2
Molecular Weight:
1041.20 (free base basis)
UNSPSC Code:
12352202
NACRES:
NA.77
assay
≥95% (HPLC)
form
powder
color
white to off-white
shipped in
wet ice
storage temp.
−20°C
Biochem/physiol Actions
45-54W (Ac-KDGIVNGVKA-NH2) is a potent peptide inhibitor of α-synuclein aggregation that reduces amyloid toxicity in cells.
45-54W is a potent peptide inhibitor of α-synuclein aggregation.
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service