Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1411

Sigma-Aldrich

BIHC

≥98% (HPLC)

Synonym(s):

3,3′-((2-Butyl-5-chloro-1H-imidazol-4-yl)methylene)bis(4-hydroxy-2H-chromen-2-one)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H21ClN2O6
CAS Number:
Molecular Weight:
492.91
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 1 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

OC1=C(C(C2=C(O)C(C=CC=C3)=C3OC2=O)C4=C(Cl)NC(CCCC)=N4)C(OC5=C1C=CC=C5)=O

InChI

1S/C26H21ClN2O6/c1-2-3-12-17-28-21(24(27)29-17)18(19-22(30)13-8-4-6-10-15(13)34-25(19)32)20-23(31)14-9-5-7-11-16(14)35-26(20)33/h4-11,18,30-31H,2-3,12H2,1H3,(H,28,29)

InChI key

YDLKVAMYKVCOFK-UHFFFAOYSA-N

Biochem/physiol Actions

BIHC is a potent TNF blocker that inhibits the proliferation of various HCC cells. BIHC exhibits potent cytotoxic activity against the HepG2 cell line while showing less toxicity towards normal hepatocytes.
BIHC is a potent TNF blocker.

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service