Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1399

Sigma-Aldrich

CCT031374 hydrobromide

≥98% (HPLC)

Synonym(s):

1-[1,1′-Biphenyl]-4-yl-2-(2,3-dihydro-9H-imidazo[1,2-a]benzimidazol-9-yl)ethanone hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H19N3O · HBr
CAS Number:
Molecular Weight:
434.33
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C1=CC=C(C2=CC=CC=C2)C=C1)CN3C4=C(C=CC=C4)N5C3=NCC5.[H]Br

InChI

1S/C23H19N3O.BrH/c27-22(19-12-10-18(11-13-19)17-6-2-1-3-7-17)16-26-21-9-5-4-8-20(21)25-15-14-24-23(25)26;/h1-13H,14-16H2;1H

InChI key

MPILENOYNNNGKO-UHFFFAOYSA-N

Biochem/physiol Actions

CCT031374 is an inhibitor of TCF-dependent transcription.
CCT031374 is an inhibitor of TCF-dependent transcription. CCT031374 is a selective β-catenin signaling inhibitor that decreases the intracellular/soluble β-cat level. CCT031374 inhibits  proliferation of numberous cancer cell lines by induction of apoptosis.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service