Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1352

Sigma-Aldrich

CINK4

≥95% (HPLC)

Synonym(s):

trans-4-[[6-(Ethylamino)-2-[[1-(phenylmethyl)-1H-indol-5-yl]amino]-4-pyrimidinyl]amino]-cyclohexanol

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$323.00

$323.00


Available to ship onApril 29, 2025Details


Request a Bulk Order

Select a Size

Change View
25 MG
$323.00

About This Item

Empirical Formula (Hill Notation):
C27H32N6O
CAS Number:
Molecular Weight:
456.58
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$323.00


Available to ship onApril 29, 2025Details


Request a Bulk Order

Quality Level

assay

≥95% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 25 mg/mL, clear

storage temp.

2-8°C

SMILES string

O[C@@H]1CC[C@@H](NC2=CC(NCC)=NC(NC3=CC=C(N(CC4=CC=CC=C4)C=C5)C5=C3)=N2)CC1

InChI

1S/C27H32N6O/c1-2-28-25-17-26(29-21-8-11-23(34)12-9-21)32-27(31-25)30-22-10-13-24-20(16-22)14-15-33(24)18-19-6-4-3-5-7-19/h3-7,10,13-17,21,23,34H,2,8-9,11-12,18H2,1H3,(H3,28,29,30,31,32)/t21-,23-

InChI key

YVXCDLCJCIDFHE-AFARHQOCSA-N

Biochem/physiol Actions

CINK4 is a specific inhbitor of CDK4/cyclin D1 complexes (IC50 = 1.5 mM). CINK4 has greater than 50 fold selectivity over CDK1 or CDK2. CINK4 dose-dependently inhibits phosphorylation of retinoblastoma (Rb) and induces G0-G1 cell cycle arrest and apoptosis in cancer cell lines.
CINK4 is a specific inhbitor of CDK4/cyclin D1 complexes.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service