Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1328

Sigma-Aldrich

AZ20

≥98% (HPLC)

Synonym(s):

4{4-[(3R)3-Methylmorpholin-4-yl]-6-[1-(methylsulfonyl)cyclopropyl]pyrimidin-2-yl}1Hindole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H24N4O3S
CAS Number:
Molecular Weight:
412.51
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

CS(C1(CC1)C2=CC(N3[C@H](C)COCC3)=NC(C4=CC=CC5=C4C=CN5)=N2)(=O)=O

InChI

1S/C21H24N4O3S/c1-14-13-28-11-10-25(14)19-12-18(21(7-8-21)29(2,26)27)23-20(24-19)16-4-3-5-17-15(16)6-9-22-17/h3-6,9,12,14,22H,7-8,10-11,13H2,1-2H3/t14-/m1/s1

InChI key

SCGCBAAYLFTIJU-CQSZACIVSA-N

Biochem/physiol Actions

AZ20 is cell penetrant potent and selective ATR (ataxia telangiectasia and Rad3-related protein) kinase inhibitor that potently inhibits the growth of LoVo colorectal adenocarcinoma xenografts in mice.
AZ20 is cell penetrant potent and selective ATR kinase inhibitor.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ilaria Stadiotti et al.
Frontiers in cardiovascular medicine, 9, 878268-878268 (2022-07-12)
Heart failure with preserved ejection fraction (HFpEF) is a heterogeneous syndrome characterized by impaired left ventricular (LV) diastolic function, with normal LV ejection fraction. Aortic valve stenosis can cause an HFpEF-like syndrome by inducing sustained pressure overload (PO) and cardiac
Madeeha Ghazi et al.
Nucleic acids research, 51(19), 10451-10466 (2023-09-12)
Melanin protects skin cells from ultraviolet radiation-induced DNA damage. However, intermediates of eumelanin are highly reactive quinones that are potentially genotoxic. In this study, we systematically investigate the effect of sustained elevation of melanogenesis and map the consequent cellular repair
Hironori Abe et al.
Nature communications, 13(1), 7212-7212 (2022-11-29)
Meiotic sex chromosome inactivation (MSCI) is an essential process in the male germline. While genetic experiments have established that the DNA damage response (DDR) pathway directs MSCI, due to limitations to the experimental systems available, mechanisms underlying MSCI remain largely

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service