Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1273

Sigma-Aldrich

ML347

≥98% (HPLC)

Synonym(s):

5-[6-(4-Methoxyphenyl)pyrazolo[1,5-a]pyrimidin-3-yl]-quinoline, LDN-193719, VU0469381

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H16N4O
CAS Number:
Molecular Weight:
352.39
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 1 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

COC(C=C1)=CC=C1C2=CN3N=CC(C4=CC=CC5=C4C=CC=N5)=C3N=C2

InChI

1S/C22H16N4O/c1-27-17-9-7-15(8-10-17)16-12-24-22-20(13-25-26(22)14-16)18-4-2-6-21-19(18)5-3-11-23-21/h2-14H,1H3

InChI key

FVRYPYDPKSZGNS-UHFFFAOYSA-N

Biochem/physiol Actions

ML347 is a derivative of dorsomorphin that is a highly selective antagonist of BMP receptor isotypes ALK1 and ALK2 (IC50 values = 46 and 32 nM, respectively). ML347 has little effect on other ALK isoforms, including ALK3 (IC50 = 10 μM).
ML347 is a highly selective antagonist of BMP receptor isotypes ALK1 and ALK2.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service