Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1262

Sigma-Aldrich

BRD7552

≥98% (HPLC)

Synonym(s):

Ethyl 4-(((((2R,3R,4S,5R,6S)-4,5-bis((benzo[d][1,3]dioxol-5-ylcarbamoyl)oxy)-2-(hydroxymethyl)-6-methoxytetrahydro-2H-pyran-3-yl)oxy)carbonyl)amino)benzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H33N3O15
CAS Number:
Molecular Weight:
711.63
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

shipped in

wet ice

storage temp.

−20°C

SMILES string

O=C(NC1=CC(OCO2)=C2C=C1)O[C@H]3[C@H](OC(NC4=CC=C(C(OCC)=O)C=C4)=O)[C@@H](CO)O[C@H](OC)[C@@H]3OC(NC5=CC(OCO6)=C6C=C5)=O

InChI

1S/C33H33N3O15/c1-3-43-29(38)17-4-6-18(7-5-17)34-31(39)49-26-25(14-37)48-30(42-2)28(51-33(41)36-20-9-11-22-24(13-20)47-16-45-22)27(26)50-32(40)35-19-8-10-21-23(12-19)46-15-44-21/h4-13,25-28,30,37H,3,14-16H2,1-2H3,(H,34,39)(H,35,40)(H,36,41)/t25-,26-,27+,28-,30+/m1/s1

InChI key

AAOLIMLEFKWKOY-IXMSMLDRSA-N

Biochem/physiol Actions

BRD7552 is a potent inducer of pancreatic and duodenal homeobox 1 (PDX1) transcription factor that enhanced insulin expression.
BRD7552 is a potent inducer of pancreatic and duodenal homeobox 1 (PDX1) transcription factor that enhanced insulin expression. BRD7552 upregulates PDX1 expression in both primary human islets and ductal cells. It appears that BRD7552 induce PDX1 expression through an epigenetic control.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service