Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1256

Sigma-Aldrich

Gyramide A hydrochloride

≥98% (HPLC)

Synonym(s):

5-Fluoro-2-methyl-N-[(3R)-1-[[4-(1-methylethoxy)phenyl]methyl]-3-pyrrolidinyl]-benzenesulfonamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27FN2O3S · HCl
CAS Number:
Molecular Weight:
442.98
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

Storage temp.

2-8°C

SMILES string

CC1=C(S(N[C@@H]2CCN(CC3=CC=C(OC(C)C)C=C3)C2)(=O)=O)C=C(F)C=C1.Cl

InChI

1S/C21H27FN2O3S.ClH/c1-15(2)27-20-8-5-17(6-9-20)13-24-11-10-19(14-24)23-28(25,26)21-12-18(22)7-4-16(21)3;/h4-9,12,15,19,23H,10-11,13-14H2,1-3H3;1H/t19-;/m1./s1

Inchi Key

LLHBXQVGBTVEDP-FSRHSHDFSA-N

Biochem/physiol Actions

Gyramide A is a selective bacterial DNA gyrase inhibitor and bacteriostatic agent with an IC50 of 3.3 ± 1.0 μM. Gyramide A competitively inhibits the ATPase activity of Escherichia coli gyrase resulting in supercoiled DNA and stopping DNA replication. Unlike other DNA gyrase inhibitors ciprofloxacin and novobiocin, Gyramide A targets only DNA gyrase without inhibiting the closely related topoisomerase IV. Gyramide A is not as potent an antibiotic, but should be a more useful tool for studying bacterial gyrase.
Gyramide A is a selective bacterial DNA gyrase inhibitor and bacteriostatic agent.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service