Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1230

Sigma-Aldrich

XW4.4

≥98% (HPLC)

Synonym(s):

5-(2,6-Dichlorophenylsulfonamido)-N-(4-fluorobenzyl)-1H-pyrrole-2-carboxamide, 5-[[(2,6-Dichlorophenyl)sulfonyl]amino]-N-[(4-fluorophenyl)methyl]-1H-pyrrole-2-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H14Cl2FN3O3S
CAS Number:
Molecular Weight:
442.29
UNSPSC Code:
12352200

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

XW4.4 can generate insulin-producing cells from rat mesenchymal stem cells. XW4.4 induced pancreatic differentiation of rat bone marrow derived mesenchymal stem cells into insulin-producing cells shown to release C-peptide and insulin in response to physiological or high glucose levels, which could lead to a possible treatment for Type 1 diabetes. Its mechanism of action is postulated to be through increasing expression of hepatocyte nuclear factor 3β (HNF3β), a member of the forkhead box transcription factor family that regulates expression of several genes in the pancreas during early embryogenesis.
XW4.4 generates insulin-producing cells from rat mesenchymal stem cells.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service