Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1219

Sigma-Aldrich

ML243

≥98% (HPLC)

Synonym(s):

(2E)-N-(4,5-Dihydro-1,3-thiazol-2-yl)-3-(4-ethylphenyl)prop-2-enamide, (2E)-N-(4,5-Dihydro-2-thiazolyl)-3-(4-ethylphenyl)-2-propenamide, (E)-N-(4,5-Dihydrothiazol-2-yl)-3-(4-ethylphenyl)acrylamide, CID 50910523, ML 243

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H16N2OS
CAS Number:
Molecular Weight:
260.35
UNSPSC Code:
12352200
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

Biochem/physiol Actions

ML243 is a potent and selective inhibitor of cancer stem-like cells (CSCs) that selectively eradicate CSCs in heterogeneous cancer cell populations.
ML243 is a potent and selective inhibitor of cancer stem-like cells (CSCs).

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service