Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1203

Sigma-Aldrich

2-PMAP

≥98% (HPLC)

Synonym(s):

(2-Pyridylmethyl)(2-hydroxyphenyl)amine, 2-N-(2-Pyridylmethyl)aminophenol, 2-[(2-Pyridinylmethyl)amino]-phenol; (2-Hydroxyphenyl)(2-pyridylmethyl)amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12N2O
CAS Number:
Molecular Weight:
200.24
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

, white to dark brown

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC1=CC=CC=C1NCC2=NC=CC=C2

InChI

1S/C12H12N2O/c15-12-7-2-1-6-11(12)14-9-10-5-3-4-8-13-10/h1-8,14-15H,9H2

InChI key

GXJRVWZNMZGWAQ-UHFFFAOYSA-N

Biochem/physiol Actions

2-PMAP is a brain penetrant inhibitor of amyloid precursor protein (APP) production.
2-PMAP is a brain penetrant inhibitor of amyloid precursor protein (APP) production. 2-PMAP lowers levels of the amyloid precursor protein (APP) in CHO APP751SW cells. Also it reduces brain level of APP in mice model. 2-PMAP exhibits no cellular toxicity up to 100 μM.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service