Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1194

Sigma-Aldrich

PT1

≥98% (HPLC)

Synonym(s):

2-Chloro-5-[[5-[[5-(4,5-dimethyl-2-nitrophenyl)-2-furanyl]methylene]-4,5-dihydro-4-oxo-2-thiazolyl]amino]-benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H16ClN3O6S
CAS Number:
Molecular Weight:
497.91
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥98% (HPLC)

form

powder

color

, yellow-red to dark brown-red

solubility

DMSO: 5 mg/mL, clear (warmed)

Storage temp.

−20°C

SMILES string

CC1=CC(C2=CC=C(O2)C=C3SC(NC4=CC(C(O)=O)=C(Cl)C=C4)=NC3=O)=C([N+]([O-])=O)C=C1C

InChI

1S/C23H16ClN3O6S/c1-11-7-16(18(27(31)32)8-12(11)2)19-6-4-14(33-19)10-20-21(28)26-23(34-20)25-13-3-5-17(24)15(9-13)22(29)30/h3-10H,1-2H3,(H,29,30)(H,25,26,28)/b20-10-

Inchi Key

RTHRCOIONCZINZ-JMIUGGIZSA-N

Biochem/physiol Actions

PT1 is a direct activator of the α-subunit of AMP-activated protein kinase (AMPK), thought to act via antagonizing the autoinhibition in the α subunits.
PT1 is a direct activator of the α-subunit of AMP-activated protein kinase (AMPK).

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service