Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1185

Sigma-Aldrich

ML338

≥98% (HPLC)

Synonym(s):

2-(4-Chloro-phenyl)-5-(1-phenyl-1H-[1,2,3]triazol-4-ylmethylsulfanyl)-[1,3,4]oxadiazole, CID 60182306

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H12ClN5OS
Molecular Weight:
369.83
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

ClC1=CC=C(C2=NN=C(SCC3=CN(C4=CC=CC=C4)N=N3)O2)C=C1

InChI

1S/C17H12ClN5OS/c18-13-8-6-12(7-9-13)16-20-21-17(24-16)25-11-14-10-23(22-19-14)15-4-2-1-3-5-15/h1-10H,11H2

InChI key

GMRWWIAYZWSOAZ-UHFFFAOYSA-N

Biochem/physiol Actions

ML338 is a inhibitor of non-replicating M. tuberculosis bacilli.
ML338 is a selective inhibitor of non-replicating Mycobacterium tuberculosis bacilli.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service