Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1156

Sigma-Aldrich

NID-1

≥98% (HPLC)

Synonym(s):

N-(2-Ethylphenyl)-3-(5-nitro-2-thienyl)- 2-propenamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H14N2O3S
CAS Number:
Molecular Weight:
302.35
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥98% (HPLC)

form

powder

color

faintly yellow to dark yellow

solubility

DMSO: 25 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NC1=CC=CC=C1CC)/C=C/C2=CC=C([N+]([O-])=O)S2

InChI

1S/C15H14N2O3S/c1-2-11-5-3-4-6-13(11)16-14(18)9-7-12-8-10-15(21-12)17(19)20/h3-10H,2H2,1H3,(H,16,18)/b9-7+

InChI key

YTQHWWVZNNZOIK-VQHVLOKHSA-N

Biochem/physiol Actions

NID-1 (Novel Inducer of Death-1) is a cell permeable and potent inducer of energy-dependent non-apoptotic cell death in mammalian cell lines. NID-1 induced cell death is distinct from macroautophagy, but still reacquires ATG5 and the lysosomal hydrolase cathepsin-L.
NID-1 is a cell permeable and potent inducer of energy-dependent non-apoptotic cell death in mammalian cell lines.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service