Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1155

Sigma-Aldrich

22-NHC

≥98% (HPLC)

Synonym(s):

(3β,20S)-20-[(3-Methylbutyl)amino]-pregn-5-en-3-ol, 22-Azacholesterol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H45NO
CAS Number:
Molecular Weight:
387.64
UNSPSC Code:
51111800

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 1 mg/mL, clear (warmed)

storage temp.

2-8°C

InChI

1S/C26H45NO/c1-17(2)12-15-27-18(3)22-8-9-23-21-7-6-19-16-20(28)10-13-25(19,4)24(21)11-14-26(22,23)5/h6,17-18,20-24,27-28H,7-16H2,1-5H3/t18-,20-,21-,22+,23-,24-,25-,26+/m0/s1

InChI key

PCLDERUMTJBTMY-MJHCCXMASA-N

Biochem/physiol Actions

22-NHC (22-Azacholesterol) is the first small molecule inhibitor that inhibits Hedgehog (Hh) signaling by binding the oxysterol-binding site of Smo. Oxysterols are the only known endogenous activators of Hedgehog signaling, with 20(S)-hydroxycholesterol being the most potent, and act at a site distinct from that targeted by current Smo inhibitors such as cyclopamine and SANT1. 22-Azacholesterol (22-NHC) inhibited Sonic Hedgehog (Shh) signaling with an IC50 of 3 μM in NIH-3T3 cells. 22-NHC binds to the same allosteric site as 20(S)-hydroxycholesterol (20-OHC), which is distinct from the cyclopamine site and the itraconazole site, and appears to act by competing with binding of 20-OHC to mSmo.
22-NHC is the first small molecule inhibitor that inhibits Hedgehog (Hh) signaling by binding the oxysterol-binding site of Smo.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service