Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1067

Sigma-Aldrich

CYM50769

≥98% (HPLC)

Synonym(s):

5-Chloro-2-(9H-fluoren-9-yl)-4-(4-methoxyphenoxy)-3(2H)-pyridazinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H17ClN2O3
CAS Number:
Molecular Weight:
416.86
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

Storage temp.

2-8°C

SMILES string

O=C1N(C2C(C=CC=C3)=C3C4=C2C=CC=C4)N=CC(Cl)=C1OC5=CC=C(OC)C=C5

InChI

1S/C24H17ClN2O3/c1-29-15-10-12-16(13-11-15)30-23-21(25)14-26-27(24(23)28)22-19-8-4-2-6-17(19)18-7-3-5-9-20(18)22/h2-14,22H,1H3

Inchi Key

QHVSQUYCVUHYKT-UHFFFAOYSA-N

Biochem/physiol Actions

CYM50769 is a potent, specific antagonist of the Neuropeptide B and Neuropeptide W receptor, NPBWR1 (IC50 = 120 nM).
CYM50769 is a potent, specific antagonist of the Neuropeptide B and Neuropeptide W receptor, NPBWR1.

Pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service