Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1056

Sigma-Aldrich

CX4338

≥98% (HPLC)

Synonym(s):

4,4-Dimethyl-6-oxo-N-phenyl-2-[(phenylmethyl)amino]-1-cyclohexene-1-carbothioamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H24N2OS
CAS Number:
Molecular Weight:
364.50
UNSPSC Code:
51111800
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

InChI

1S/C22H24N2OS/c1-22(2)13-18(23-15-16-9-5-3-6-10-16)20(19(25)14-22)21(26)24-17-11-7-4-8-12-17/h3-12,23H,13-15H2,1-2H3,(H,24,26)

InChI key

AESCWMGFNWXKFR-UHFFFAOYSA-N

Biochem/physiol Actions

CX4338 is a CXCR2 receptor antagonist.
CX4338 is a CXCR2 receptor antagonist. In cell based assays, CX4338 blocks CXCL8-induced receptor internalization and β-arrestin recruitment, but enhances G-protein-mediated signaling events such as calcium mobilization and ERK phosporylation. The comopund inhibits CXCL8-induced neutrophil chemotaxis, and LPS-induced PMN migration in a mouse airway inflammaiton model.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service