Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1054

Sigma-Aldrich

SBI-0087702

≥98% (HPLC)

Synonym(s):

4-Methoxy-N-[2-(4-methoxyphenyl)ethyl]-1-naphthalenemethanamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H23NO2
CAS Number:
Molecular Weight:
321.41
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 5 mg/mL, clear (warmed)

Storage temp.

room temp

SMILES string

COC1=C2C(C=CC=C2)=C(CNCCC3=CC=C(OC)C=C3)C=C1

InChI

1S/C21H23NO2/c1-23-18-10-7-16(8-11-18)13-14-22-15-17-9-12-21(24-2)20-6-4-3-5-19(17)20/h3-12,22H,13-15H2,1-2H3

Inchi Key

SNBZXHDTILHVJS-UHFFFAOYSA-N

Biochem/physiol Actions

SBI-0087702 promotes the cytoplasmic localization of ATF2 in melanoma cells. SBI-0087702-induced translocation of ATF2 to the mitochondria results in increased apoptosis due to loss of mitochondrial membrane integrity. SBI-0087702 also inhibits growth and motility of melanoma cells. SBI-0087702 was shown to inhibit ATF2 phosphorylation on Thr52 by PKCε.
SBI-0087702 promotes the cytoplasmic localization of ATF2, which induces apoptosis and inhibits growth and motility of melanoma cells.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

CorrosionEnvironment

signalword

Danger

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service