SML1040
SCH-50911
≥95% (HPLC)
Synonym(s):
(+)-(2S)-5,5-Dimethyl-2-morpholineacetic acid hydrochloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C8H15NO3 · HCl
CAS Number:
Molecular Weight:
209.67
UNSPSC Code:
12352200
assay
≥95% (HPLC)
form
oil
optical activity
[α]/D +12 to +20°, c = 1 in H2O
storage condition
desiccated
color
white to beige
solubility
H2O: 25 mg/mL, clear
Storage temp.
−20°C
Biochem/physiol Actions
SCH-50911 is a GABAB receptor antagonist.
SCH-50911 is a selective, competitive, orally active GABAB receptor antagonist.
Features and Benefits
This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service