Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1036

Sigma-Aldrich

LDN-193594

≥98% (HPLC)

Synonym(s):

[4-Amino-2-(3-pyridinylamino)-5-thiazolyl](2-fluorophenyl)-methanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11FN4OS
CAS Number:
Molecular Weight:
314.34
UNSPSC Code:
12352200
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

InChI

1S/C15H11FN4OS/c16-11-6-2-1-5-10(11)12(21)13-14(17)20-15(22-13)19-9-4-3-7-18-8-9/h1-8H,17H2,(H,19,20)

InChI key

XHJMMMPXTUEBMQ-UHFFFAOYSA-N

Biochem/physiol Actions

LDN-193594 is a potent and nontoxic dual inhibitor of CDK5/p25 and GSK3β that reduces tau protein phosphorylation and exerts neuroprotective effects in vivo. LDN-193594 improves the tauopathy in mice models.
LDN-193594 is a potent and nontoxic dual inhibitor of CDK5/p25 and GSK3β.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service