Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1028

Sigma-Aldrich

MPT0B014

≥98% (HPLC)

Synonym(s):

6-Quinolinyl(3,4,5-trimethoxyphenyl)methanone, B014

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H17NO4
CAS Number:
Molecular Weight:
323.34
UNSPSC Code:
12352200
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

Biochem/physiol Actions

MPT0B014 inhibits tubulin polymerization and disrupts microtubules in the cytoskeleton.
MPT0B014 inhibits tubulin polymerization and disrupts microtubules in the cytoskeleton. The compound MPT0B014 inhibits proliferation of the lung cancer cells A549, H1299, and H226 with IC50 values of 41, 46 and 42 nM, respectively. MPT0B014 induces cell cycle arrest and apoptosis, inhibits tumor growth in lung cancer xenografts, and potentiates inhibition of tumor growth by erlotinib.

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service