Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1024

Sigma-Aldrich

WHI-P154

≥98% (HPLC)

Synonym(s):

2-Bromo-4-[(6,7-dimethoxy-4-quinazolinyl)amino]-phenol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14BrN3O3
CAS Number:
Molecular Weight:
376.20
UNSPSC Code:
12352200
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to brown

solubility

DMSO: 1 mg/mL, clear (warmed)

storage temp.

−20°C

InChI

1S/C16H14BrN3O3/c1-22-14-6-10-12(7-15(14)23-2)18-8-19-16(10)20-9-3-4-13(21)11(17)5-9/h3-8,21H,1-2H3,(H,18,19,20)

InChI key

CBIAKDAYHRWZCU-UHFFFAOYSA-N

Biochem/physiol Actions

WHI-P154 is a specific inhibitor of JAK3 (IC50 = 1.8 μM) with no inhibitory activity against JAK1 or JAK2. WHI-P154 enhances myogenic differentiation of C2C12 mouse myoblast cells, and induces differentiation of neural progenitor cells. The compound WHI-P154 dose dependently inhibits expression of iNOS and NO production in LPS-challenged macrophages.
WHI-P154 is a specific inhibitor of JAK3.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service