Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1015

Sigma-Aldrich

OS-3-106

≥98% (HPLC)

Synonym(s):

N-(4-(4-(2-Methoxyphenyl)piperazin-1-yl)butyl)-4-(thiazol-4-yl)benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H30N4O2S
CAS Number:
Molecular Weight:
450.60
UNSPSC Code:
12352200
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

OS-3-106 is a potent and selective dopamine D3 receptor (D3R) partial agonist that blocked a D3R-mediated behavior in rats. OS-3-106 is effective in reducing cocaine self-administration.
OS-3-106 is a potent and selective dopamine D3 receptor (D3R) partial agonist.

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service