Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0988

Sigma-Aldrich

CCCI-01

≥98% (HPLC)

Synonym(s):

5-Nitro-N-(3-pyridinylmethyl)-2-furancarboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$97.85

$97.85

List Price$103.00Save 5%
Web-Only Promotion

Available to ship onApril 28, 2025Details


Request a Bulk Order

Select a Size

Change View
5 MG
$97.85

About This Item

Empirical Formula (Hill Notation):
C11H9N3O4
CAS Number:
Molecular Weight:
247.21
UNSPSC Code:
12352200
NACRES:
NA.77

$97.85

List Price$103.00Save 5%
Web-Only Promotion

Available to ship onApril 28, 2025Details


Request a Bulk Order

Quality Level

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

[N+](=O)([O-])c1[o]c(cc1)C(=O)NCc2cnccc2

InChI

1S/C11H9N3O4/c15-11(9-3-4-10(18-9)14(16)17)13-7-8-2-1-5-12-6-8/h1-6H,7H2,(H,13,15)

InChI key

OBZHOVWNZIVCCZ-UHFFFAOYSA-N

Biochem/physiol Actions

CCCI-01 iinduces the fomation of multipolar spindles in cancer cells.
CCCI-01 induces the fomation of multipolar spindles in cancer cells containing multiple, or supernumerary centromeres. CCCI-01 leads to centromere declustering, disruption of cell cycle and apoptsis in BT-549 breast cancer cells without affecting bipolar spindles in normal primary human mammary epithelial cells.

Features and Benefits

This compound is a featured product for Apoptosis research. Click here to discover more featured Apoptosis products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Apoptosis regulation involves multiple pathways and molecules for cellular homeostasis.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service