Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0973

Sigma-Aldrich

NSC 37249 dihydrochloride

≥98% (HPLC)

Synonym(s):

N,N′-(Sulfonyldi-4,1-phenylene)bis[2-(dicyclohexylamino)-acetamide dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H58N4O4S · 2HCl
CAS Number:
Molecular Weight:
763.90
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=S(C(C=C1)=CC=C1NC(CN(C2CCCCC2)C3CCCCC3)=O)(C4=CC=C(NC(CN(C5CCCCC5)C6CCCCC6)=O)C=C4)=O.Cl.Cl

InChI

1S/C40H58N4O4S.2ClH/c45-39(29-43(33-13-5-1-6-14-33)34-15-7-2-8-16-34)41-31-21-25-37(26-22-31)49(47,48)38-27-23-32(24-28-38)42-40(46)30-44(35-17-9-3-10-18-35)36-19-11-4-12-20-36;;/h21-28,33-36H,1-20,29-30H2,(H,41,45)(H,42,46);2*1H

InChI key

MYOCKQZXPRANOY-UHFFFAOYSA-N

Biochem/physiol Actions

NSC 37249 is a potent inhibitor of adenovirus proteinase (IC50 = 0.14 μM).
NSC 37249 is a potent inhibitor of adenovirus proteinase.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service