Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0969

Sigma-Aldrich

Inauhzin

≥98% (HPLC)

Synonym(s):

1-(10H-Phenothiazin-10-yl)-2-(5H-1,2,4-triazino[5,6-b]indol-3-ylthio)-1-butanone, 10-[2-(5H-[1,2,4]Triazino[5,6-b]indol-3-ylthio)butanoyl]-10H-phenothiazine, INZ

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H19N5OS2
CAS Number:
Molecular Weight:
469.58
UNSPSC Code:
12352200

assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

InChI

1S/C25H19N5OS2/c1-2-19(33-25-27-23-22(28-29-25)15-9-3-4-10-16(15)26-23)24(31)30-17-11-5-7-13-20(17)32-21-14-8-6-12-18(21)30/h3-14,19H,2H2,1H3,(H,26,27,29)

InChI key

VHUOXERIKQWIJE-UHFFFAOYSA-N

Biochem/physiol Actions

Inauhzin is a cell-permeable, selective inhibitor of SirT1, which effectively reactivates p53 by inhibiting SIRT1 activity. Inauhzin inhibits cell proliferation, induces senescence, and p53-dependent apoptosis of human cancer cells without apparent toxicity to normal cells.
Inauhzin is a cell-permeable, selective inhibitor of SirT1; p53 activator.

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service