Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0966

Sigma-Aldrich

PLX647

≥98% (HPLC)

Synonym(s):

5-(1H-Pyrrolo[2,3-b]pyridin-3-ylmethyl)-N-[[4-(trifluoromethyl)phenyl]methyl]-2-pyridinamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H17F3N4
CAS Number:
Molecular Weight:
382.38
UNSPSC Code:
12352200
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

Storage temp.

2-8°C

Biochem/physiol Actions

PLX647 is a potent and specific dual inhibitor of the receptor tyrosine kinases FMS and KIT.
PLX647 is a potent and specific dual inhibitor of the receptor tyrosine kinases FMS and KIT. PLX647 blocks activation of macrophages, osteoclasts and mast cells in both cell based and in vivo inflammation models.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service