Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0938

Sigma-Aldrich

Dofequidar fumarate

≥98% (HPLC)

Synonym(s):

4-(Diphenylacetyl)-a-[(5-quinolinyloxy)methyl]-1-Piperazineethanol (E)-2-butenedioate fumarate (1:1.5), MS-209 fumarate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H31N3O3 · 1.5 C4H4O4
CAS Number:
Molecular Weight:
655.69
UNSPSC Code:
12161501
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

InChI

1S/2C30H31N3O3.3C4H4O4/c2*34-25(22-36-28-15-7-14-27-26(28)13-8-16-31-27)21-32-17-19-33(20-18-32)30(35)29(23-9-3-1-4-10-23)24-11-5-2-6-12-24;3*5-3(6)1-2-4(7)8/h2*1-16,25,29,34H,17-22H2;3*1-2H,(H,5,6)(H,7,8)/b;;3*2-1+

InChI key

ZGMJYTYLTJFNCS-VQYXCCSOSA-N

Biochem/physiol Actions

Dofequidar fumarate is a potent inhibitor of MDR-1.
Dofequidar fumarate is a potent inhibitor of MDR-1. The compound Dofequidar fumarate sensitizes tumor cells to cytotoxic drugs.

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Discover Bioactive Small Molecules for ADME/Tox

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service