Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0926

Sigma-Aldrich

ML314 maleate

≥98% (HPLC)

Synonym(s):

2-Cyclopropyl-6,7-dimethoxy-4-(4-(2-methoxyphenyl)-piperazin-1-yl)quinazoline maleate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$101.00

$101.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$101.00

About This Item

Empirical Formula (Hill Notation):
C24H28N4O3 · C4H4O4
Molecular Weight:
536.58
UNSPSC Code:
12352202
NACRES:
NA.77

$101.00


Check Cart for Availability

Request a Bulk Order

Quality Level

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

Biochem/physiol Actions

ML314 is a biased agonist of the Neruotensin 1 (NTR1) receptor. ML314 induces recruitment of β-arrestin to NTR1 with an EC50 of 2 μM, but does not induce G-protein mediated Ca2+ mobilization.
ML314 is a neurotensin receptor (NTR1) receptor.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service