Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0894

Sigma-Aldrich

VU0420373

≥98% (HPLC)

Synonym(s):

2-[5-(2-Fluorophenyl)-1H-pyrazol-3-yl]-phenol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11FN2O
CAS Number:
Molecular Weight:
254.26
UNSPSC Code:
12352200
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mg protein, clear

storage temp.

2-8°C

Biochem/physiol Actions

VU0420373 is an activator of the heme sensor system (HssRS) in S. aureus.
VU0420373 is an activator of the heme sensor system (HssRS) in Staphylococcus aureus. The compound VU0420373 induces heme biosynthesis, and is toxic to fermenting S. aureus. VU0420373 reduces the number of liver abscesses, and tissue damage in S. aureus infected mice.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service