Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0890

Sigma-Aldrich

SB 452533

≥98% (HPLC)

Synonym(s):

N-(2-Bromophenyl)-N′-[2-[ethyl(3-methylphenyl)amino]ethyl]urea, SB-452533

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H22BrN3O
CAS Number:
Molecular Weight:
376.29
UNSPSC Code:
12352200
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

yellow to orange

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

InChI

1S/C18H22BrN3O/c1-3-22(15-8-6-7-14(2)13-15)12-11-20-18(23)21-17-10-5-4-9-16(17)19/h4-10,13H,3,11-12H2,1-2H3,(H2,20,21,23)

InChI key

IFJYEGJUQIBBQV-UHFFFAOYSA-N

Biochem/physiol Actions

SB-452533 acts as a competitive inhibitor at the capsaicin binding site on the transient receptor potential channel TRPV1 receptor with a pKb of 7.7 versus capsaicin and pIC50 of 7.0 versus pH. At higher concentrations, SB-452533 is also active at TRPM8 with an IC50 of 571 nM. It has a high clearance rate, so is not suitable for in vivo use.
SB-452533 acts as a competitive inhibitor at the capsaicin binding site on the transient receptor potential channel TRPV1 receptor.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

DISCOVER Bioactive Small Molecules for Neuroscience

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service