SML0880
Shz-1
≥98% (HPLC)
Synonym(s):
2-[(5-Bromo-2-hydroxyphenyl)methylene]hydrazide benzenesulfonic acid
About This Item
assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 10 mg/mL, clear
Storage temp.
−20°C
InChI
1S/C13H11BrN2O3S/c14-11-6-7-13(17)10(8-11)9-15-16-20(18,19)12-4-2-1-3-5-12/h1-9,16-17H/b15-9+
Inchi Key
OEMCLQLAWYKPRK-OQLLNIDSSA-N
Biochem/physiol Actions
Features and Benefits
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service