SML0862
RX871024
≥98% (HPLC)
Synonym(s):
2-(4,5-Dihydro-1H-imidazol-2-yl)-1-phenyl-1H-indole hydrochloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C17H15N3 · HCl
CAS Number:
Molecular Weight:
297.78
UNSPSC Code:
12352200
NACRES:
NA.77
assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
H2O: 15 mg/mL, clear
Storage temp.
2-8°C
Biochem/physiol Actions
RX871024 is an imidazoline I3 receptor antagonist.
RX871024 is an imidazoline receptor antagonist that stimulates insulin secretion in β-cells and islets at lower concentrations than other imidazoline ligands. 0.1 μM RX871024 enhances glucose-induced insulin secretion under nonpolarizing conditions and at higher concentrations, it enhances secretion through a KATP channel-independent mechanism.
Features and Benefits
This compound is featured on the Imidazoline Binding Sites page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service