Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0855

Sigma-Aldrich

GW438014A

≥98% (HPLC)

Synonym(s):

GW 438014A, N-[1-(2-Phenylethyl)-1H-benzimidazol-2-yl]-benzamide monomethanesulfonate (9CI)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H19N3O · CH3SO3H
CAS Number:
Molecular Weight:
437.51
UNSPSC Code:
12352200

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

InChI

1S/C22H19N3O.CH4O3S/c26-21(18-11-5-2-6-12-18)24-22-23-19-13-7-8-14-20(19)25(22)16-15-17-9-3-1-4-10-17;1-5(2,3)4/h1-14H,15-16H2,(H,23,24,26);1H3,(H,2,3,4)

InChI key

AVYFZOKRFCFQLP-UHFFFAOYSA-N

Biochem/physiol Actions

GW438014A is a potent and selective neuropeptide Y Y5 receptor antagonist.
GW438014A is a potent and selective neuropeptide Y Y5 receptor antagonist. GW438014A induces a potent reduction of food intake in rodents.

Features and Benefits

This compound is featured on the Neuropeptide Y Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service